3-Formyl-6-methylchromone 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Methyl-4-oxo-4H-chromene-3-carbaldehyde
A139494-1G
A139494-250MG
A139494-5G
76767-706EA
11.67
USD
76767-706
76767-708
76767-710
3-Formyl-6-methylchromone 95%
3-Formyl-6-methylchromone
Precaución: for Research Use Only
Formula:
C₁₁H₈O₃ MW: 188.1794 g/mol |
MDL Number:
MFCD00138943 CAS Number: 42059-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...