3-(Pyridin-4-yl)propan-1-amine 97%

Supplier: Ambeed
Warning

A139358-1G A139358-5G A139358-250MG
76767-700EA 140.04 USD
76767-700 76767-702 77566-794
3-(Pyridin-4-yl)propan-1-amine 97%
3-(Pyridin-4-yl)propan-1-amine

Precaución: For research use only.

Formula: C₈H₁₂N₂
MDL Number: MFCD08234495
CAS Number: 30532-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR