L-Glutamic acid dibenzyl ester hydrochloride 98%

Supplier: Ambeed
Warning

A138983-1G A138983-5G A138983-100G A138983-25G A138983-500G
76767-688EA 14.99 USD
76767-688 76767-690 77597-110 77597-112 77876-967
L-Glutamic acid dibenzyl ester hydrochloride 98%
L-Glutamic acid dibenzyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₂₁NO₄·HCl
MDL Number: MFCD00035103
CAS Number: 4561-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR