5-((tert-Butoxycarbonyl)amino)nicotinic acid 97%

Supplier: Ambeed
Warning

A138692-100MG A138692-1G A138692-250MG
76767-682EA 14 USD
76767-682 76767-684 76767-686
5-((tert-Butoxycarbonyl)amino)nicotinic acid 97%
5-((tert-Butoxycarbonyl)amino)nicotinic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₄
MW: 238.23 g/mol
MDL Number: MFCD06657509
CAS Number: 337904-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR