Ethyl-2-aminothiazole-4-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-aminothiazole-4-carboxylate
A138538-100G
A138538-10G
A138538-25G
A138538-500G
A138538-5G
76767-670EA
60
USD
76767-670
76767-672
76767-674
76767-676
76767-678
Ethyl-2-aminothiazole-4-carboxylate 98%
Ethyl-2-aminothiazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₂S |
MDL Number:
MFCD00619079 CAS Number: 5398-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...