5,6-Dichloro-1H-1,3-benzimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichlorobenzimidazole, 5,6-Dichloro-1H-benzo[d]imidazole

A138424-100MG A138424-1G A138424-250MG A138424-5G A138424-25G
76767-652EA 15.92 USD
76767-652 76767-654 76767-656 76767-658 77624-154
5,6-Dichloro-1H-1,3-benzimidazole 97%
5,6-Dichloro-1H-1,3-benzimidazole

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
MDL Number: MFCD00211325
CAS Number: 6478-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR