5-Hydroxy-2-adamantanone 98%

Supplier: Ambeed
Warning

Synonyms: Idramantone, 5-Hydroxyadamantan-2-one

A138321-100G A138321-100MG A138321-10G A138321-1G A138321-25G A138321-5G A138321-500G
76767-634EA 131.56 USD
76767-634 76767-636 76767-638 76767-640 76767-642 76767-644 77597-096
5-Hydroxy-2-adamantanone 98%
5-Hydroxy-2-adamantanone

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
MDL Number: MFCD00192211
CAS Number: 20098-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR