1-Bromo-3,5-bis(trifluoromethyl)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Bis(trifluoromethyl)-1-bromobenzene, 1-Bromo-3,5-bis(trifluoromethyl)benzene, 5-Bromo-α,α,α,β,β,β-Hexafluoro-m-xylene, 1,3-Bis(trifluoromethyl)-5-bromobenzene, 3,5-Bis(trifluoromethyl)bromobenzene, 5-Bromo-α,α,α,α',α',α'-Hexafluoro-m-xylene

A137022-100G A137022-10G A137022-1000G A137022-500G
76767-590EA 18 USD
76767-590 76767-592 77566-724 77566-726
1-Bromo-3,5-bis(trifluoromethyl)benzene 98%
1-Bromo-3,5-bis(trifluoromethyl)benzene

Precaución: For research use only.

Formula: C₈H₃BrF₆
MDL Number: MFCD00000381
CAS Number: 328-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR