Cyclohexylacetic acid 97%

Supplier: Ambeed
Warning

A136975-10G A136975-1G A136975-25G A136975-5G A136975-100G
76767-582EA 15 USD
76767-582 76767-584 76767-586 76767-588 77599-992
Cyclohexylacetic acid 97%
Cyclohexylacetic acid

Precaución: for Research Use Only

Formula: C₈H₁₄O₂
MDL Number: MFCD00001518
CAS Number: 5292-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR