2,3,5-Trimethylpyrazine 97%

Supplier: Ambeed
Danger

A136805-10G A136805-5G A136805-25G
76767-578EA 6.37 USD
76767-578 76767-580 77315-270
2,3,5-Trimethylpyrazine 97%
2,3,5-Trimethylpyrazine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00006145
CAS Number: 14667-55-1
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR