2-Bromo-4-fluoro-6-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-α,α,α,4-tetrafluoro-o-toluidine

A136407-1G A136407-25G A136407-5G A136407-100G
76767-532EA 23 USD
76767-532 76767-534 76767-536 77876-955
2-Bromo-4-fluoro-6-(trifluoromethyl)aniline 98%
2-Bromo-4-fluoro-6-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄BrF₄N
MW: 258.01
MDL Number: MFCD04973756
CAS Number: 875664-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR