5-Amino-2-methylbenzenesulfonamide 95%

Supplier: Ambeed
Warning

A135739-10G A135739-1G A135739-25G A135739-5G
76767-466EA 9.55 USD
76767-466 76767-468 76767-470 76767-472
5-Amino-2-methylbenzenesulfonamide 95%
5-Amino-2-methylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
MDL Number: MFCD06681071
CAS Number: 6973-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR