Methyl thiosalicylate 97%

Supplier: Ambeed
Danger

Synonyms: Methyl 2-mercaptobenzoate

A135282-100G A135282-10G A135282-1G A135282-5G A135282-25G
76767-432EA 235 USD
76767-432 76767-434 76767-436 76767-438 77549-634
Methyl thiosalicylate 97%
Methyl thiosalicylate

Precaución: for Research Use Only

Formula: C₈H₈O₂S
MDL Number: MFCD00060678
CAS Number: 4892-02-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR