(Hydroxymethyl)triphenylphosphonium chloride 95%

Supplier: Ambeed
Warning

A134969-100G A134969-10G A134969-1G A134969-25G A134969-5G
76767-396EA 267.35 USD
76767-396 76767-398 76767-400 76767-402 76767-404
(Hydroxymethyl)triphenylphosphonium chloride 95%
(Hydroxymethyl)triphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₁₉H₁₈ClOP
MW: 328.7724 g/mol
MDL Number: MFCD00031688
CAS Number: 5293-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR