5'-Chloro-5'-deoxyadenosine 97%

Supplier: Ambeed
Warning

A134694-10G A134694-1G A134694-250MG A134694-5G A134694-25G
76767-388EA 42.44 USD
76767-388 76767-390 76767-392 76767-394 77566-642
5'-Chloro-5'-deoxyadenosine 97%
5'-Chloro-5'-deoxyadenosine

Precaución: For research use only.

Formula: C₁₀H₁₂ClN₅O₃
MW: 285.69 g/mol
MDL Number: MFCD00049038
CAS Number: 892-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR