5-Chloroisoindolin-1-one 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2,3-dihydro-1-oxo-1H-isoindole, 5-Chloro-2,3-dihydro-1H-isoindol-1-one

A134539-100MG A134539-10G A134539-1G A134539-250MG A134539-5G
76767-378EA 29.71 USD
76767-378 76767-380 76767-382 76767-384 76767-386
5-Chloroisoindolin-1-one 98%
5-Chloroisoindolin-1-one

Precaución: for Research Use Only

Formula: C₈H₆ClNO
MW: 167.592 g/mol
MDL Number: MFCD08234731
CAS Number: 74572-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR