5-Bromo-2-chloro-3-methoxypyridine 98%

Supplier: Ambeed
Warning

A134273-100MG A134273-1G A134273-250MG A134273-25G A134273-5G
76767-366EA 6.37 USD
76767-366 76767-368 76767-370 76767-372 76767-374
5-Bromo-2-chloro-3-methoxypyridine 98%
5-Bromo-2-chloro-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MDL Number: MFCD08234976
CAS Number: 286947-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR