2-Fluoro-4-nitroanisole 98%

Supplier: Ambeed
Warning

A134220-1G A134220-25G
76767-362EA 5.31 USD
76767-362 76767-364
2-Fluoro-4-nitroanisole 98%
2-Fluoro-4-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MW: 171.13 g/mol
MDL Number: MFCD00061095
CAS Number: 455-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR