3-Fluoro-4-iodobenzaldehyde 97%

Supplier: Ambeed
Warning

A131967-10G A131967-1G A131967-250MG
76767-276EA 458.31 USD
76767-276 76767-278 76767-280
3-Fluoro-4-iodobenzaldehyde 97%
3-Fluoro-4-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C7H4FIO
MW: 250.0100 g/mol
MDL Number: MFCD07782055
CAS Number: 1003709-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR