6-Bromo-1,1,4,4,7-pentamethyl-1,2,3,4-tetrahydronaphthalene 95%

Supplier: Ambeed
Warning

A131716-100G A131716-10G A131716-1G A131716-25G A131716-5G
76767-266EA 531.52 USD
76767-266 76767-268 76767-270 76767-272 76767-274
6-Bromo-1,1,4,4,7-pentamethyl-1,2,3,4-tetrahydronaphthalene 95%
6-Bromo-1,1,4,4,7-pentamethyl-1,2,3,4-tetrahydronaphthalene

Precaución: for Research Use Only

Formula: C₁₅H₂₁Br
MW: 281.231 g/mol
MDL Number: MFCD08235072
CAS Number: 119999-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR