2-Chloro-6-fluorobenzaldehyde 97% (by GC)

Supplier: Ambeed
Warning

A131674-1000G A131674-100G A131674-500G A131674-25G
76767-260EA 134.74 USD
76767-260 76767-262 76767-264 77314-126
2-Chloro-6-fluorobenzaldehyde 97% (by GC)
2-Chloro-6-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClFO
MW: 158.56 g/mol
MDL Number: MFCD00003306
CAS Number: 387-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR