6-Methoxy-2-naphthaldehyde 95%

Supplier: Ambeed
Warning

A131663-100G A131663-10G A131663-25G A131663-500G A131663-5G
76767-250EA 49.87 USD
76767-250 76767-252 76767-254 76767-256 76767-258
6-Methoxy-2-naphthaldehyde 95%
6-Methoxy-2-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
MDL Number: MFCD00081152
CAS Number: 3453-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR