Ethyl-1,3-dimethylpyrazole-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1,3-dimethyl-1H-pyrazole-5-carboxylate, 1,3-Dimethylpyrazole-5-carboxylic Acid Ethyl Ester

A130792-100G A130792-10G A130792-1G A130792-25G A130792-5G
76767-220EA 213.25 USD
76767-220 76767-222 76767-224 76767-226 76767-228
Ethyl-1,3-dimethylpyrazole-5-carboxylate 97%
Ethyl-1,3-dimethylpyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MW: 168.20 g/mol
MDL Number: MFCD00233452
CAS Number: 5744-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR