Ethyl-5-methyl-1H-pyrrole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-methyl-1H-pyrrole-2-carboxylate

A130634-1G A130634-250MG A130634-10G A130634-5G A130634-100G A130634-25G
76767-208EA 24.41 USD
76767-208 76767-210 77592-508 77592-510 77596-718 77596-720
Ethyl-5-methyl-1H-pyrrole-2-carboxylate 98%
Ethyl-5-methyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MW: 153.1800 g/mol
MDL Number: MFCD01006731
CAS Number: 3284-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR