5-Bromo-1-naphthoic acid 98%

Supplier: Ambeed
Warning

A130338-100G A130338-10G A130338-1G A130338-250MG A130338-25G A130338-5G
76767-192EA 559 USD
76767-192 76767-194 76767-196 76767-198 76767-200 76767-202
5-Bromo-1-naphthoic acid 98%
5-Bromo-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₇BrO₂
MDL Number: MFCD00092713
CAS Number: 16726-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR