2-Amino-6-phenylpyrimidin-4(3H)-one 97%

Supplier: Ambeed
Warning

A130169-1G A130169-250MG A130169-5G A130169-25G
76767-180EA 18 USD
76767-180 76767-182 76767-184 77876-938
2-Amino-6-phenylpyrimidin-4(3H)-one 97%
2-Amino-6-phenylpyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₁₀H₉N₃O
MDL Number: MFCD00466412
CAS Number: 56741-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR