4,5-Difluoroanthranilic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-4,5-difluorobenzoic acid
A130114-100G
A130114-10G
A130114-1G
A130114-25G
A130114-5G
76767-166EA
159.14
USD
76767-166
76767-168
76767-170
76767-172
76767-174
4,5-Difluoroanthranilic acid 97%
4,5-Difluoroanthranilic acid
Precaución: for Research Use Only
Formula:
C₇H₅F₂NO₂ MW: 173.12 g/mol |
MDL Number:
MFCD00077509 CAS Number: 83506-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...