5-Chloro-6-methoxyindole 95%

Supplier: Ambeed
Warning

A130053-250MG A130053-1G A130053-5G A130053-100MG
76767-164EA 26 USD
76767-164 77624-100 77624-102 77876-937
5-Chloro-6-methoxyindole 95%
5-Chloro-6-methoxyindole

Precaución: for Research Use Only

Formula: C₉H₈ClNO
MDL Number: MFCD04972054
CAS Number: 90721-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR