6-(Dimethylamino)-2-naphthaldehyde 95%

Supplier: Ambeed
Warning

A130030-100MG A130030-1G A130030-250MG A130030-5G
76767-156EA 33.95 USD
76767-156 76767-158 76767-160 76767-162
6-(Dimethylamino)-2-naphthaldehyde 95%
6-(Dimethylamino)-2-naphthaldehyde

Precaución: for Research Use Only

Formula: C13H13NO
MW: 199.2484 g/mol
MDL Number: MFCD07776686
CAS Number: 173471-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR