1-Phenyl-2-pentanone 95%

Supplier: Ambeed
Warning

Synonyms: 1-Phenylpentan-2-one

A129581-1G A129581-250MG
76767-130EA 60.48 USD
76767-130 77624-098
1-Phenyl-2-pentanone 95%
1-Phenyl-2-pentanone

Precaución: for Research Use Only

Formula: (C₆H₅)CH₂C(O)(CH₂)₂CH₃
MW: 162.23 g/mol
MDL Number: MFCD00027142
CAS Number: 6683-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR