3-(4-Fluorobenzoyl)propionic acid 97%

Supplier: Ambeed
Warning

A129486-100G A129486-10G A129486-25G A129486-500G A129486-5G
76767-120EA 95 USD
76767-120 76767-122 76767-124 76767-126 76767-128
3-(4-Fluorobenzoyl)propionic acid 97%
3-(4-Fluorobenzoyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₉FO₃
MDL Number: MFCD00002793
CAS Number: 366-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR