1,5-Dimethyl-1H-pyrazole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A129307-100G A129307-10G A129307-1G A129307-25G
76767-100EA 252.5 USD
76767-100 76767-102 76767-104 76767-106
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid 98%
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD00085040
CAS Number: 5744-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR