6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene 97%

Supplier: Ambeed
Warning

A129290-10G A129290-1G A129290-250MG A129290-25G A129290-5G
76767-092EA 153.84 USD
76767-092 76767-094 76767-096 76767-098 77566-514
6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene 97%
6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene

Precaución: For research use only.

Formula: C₁₄H₁₉Br
MDL Number: MFCD05664407
CAS Number: 27452-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR