5-Nitro-2-piperidinobenzenecarbonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 5-Nitro-2-piperidin-1-ylbenzonitrile

A128951-250MG A128951-5G
76767-076EA 10.61 USD
76767-076 76767-078
5-Nitro-2-piperidinobenzenecarbonitrile 95%
5-Nitro-2-piperidinobenzenecarbonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₃N₃O₂
MW: 231.26 g/mol
MDL Number: MFCD05365582
CAS Number: 32188-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR