2-Fluoro-5-nitrobenzotrifluoride 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-nitrobenzotrifluoride, 1-Fluoro-4-nitro-2-(trifluoromethyl)benzene

A128707-25G A128707-500G A128707-100G
76767-070EA 16 USD
76767-070 76767-072 77876-928
2-Fluoro-5-nitrobenzotrifluoride 98%
2-Fluoro-5-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃F₄NO₂
MDL Number: MFCD00024576
CAS Number: 400-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR