3-Amino-4-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

A128662-10G A128662-1G A128662-25G A128662-5G
76767-062EA 25 USD
76767-062 76767-064 76767-066 76767-068
3-Amino-4-fluorobenzoic acid 97%
3-Amino-4-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD00130055
CAS Number: 2365-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR