Ethyl-2-(2-methyl-1,3-dioxolan-2-yl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(2-methyl-1,3-dioxolan-2-yl)acetate, 2-(2-Methyl-1,3-dioxolan-2-yl)acetic Acid Ethyl Ester

A128585-1000G A128585-100G A128585-10G A128585-500G A128585-25G
76767-044EA 63.66 USD
76767-044 76767-046 76767-048 76767-050 77313-494
Ethyl-2-(2-methyl-1,3-dioxolan-2-yl)acetate 98%
Ethyl-2-(2-methyl-1,3-dioxolan-2-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₁₄O₄
MW: 174.20 g/mol
MDL Number: MFCD00152488
CAS Number: 6413-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR