5-Iodonicotinic acid 95%

Supplier: Ambeed
Warning

A128536-100MG A128536-1G A128536-250MG
76767-032EA 45 USD
76767-032 76767-034 76767-036
5-Iodonicotinic acid 95%
5-Iodonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄INO₂
MDL Number: MFCD07437961
CAS Number: 15366-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR