4-Chloro-3-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2-chloro-α,α,α-trifluorotoluene, 1-Amino-4-chloro-3-(trifluoromethyl)benzene, 4-Chloro-α,α,α-trifluoro-m-toluidine, 5-Amino-2-chlorobenzotrifluoride

A128521-100G A128521-10G A128521-500G A128521-25G A128521-1000G
76767-026EA 43 USD
76767-026 76767-028 76767-030 77313-472 77624-092
4-Chloro-3-(trifluoromethyl)aniline 98%
4-Chloro-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅ClF₃N
MW: 195.57 g/mol
MDL Number: MFCD00007846
CAS Number: 320-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR