Brilliant yellow ≥85% (dye content)

Supplier: Ambeed
Warning

Synonyms: Sodium 6,6'-(ethene-1,2-diyl)bis(3-((4-hydroxyphenyl)diazenyl)benzenesulphonate), Direct Yellow 4

A128436-25G A128436-5G A128436-100G
76767-024EA 35 USD
76767-024 77596-712 77876-923
Brilliant yellow ≥85% (dye content)
Brilliant yellow

Precaución: for Research Use Only

Formula: C₂₆H₂₀N₄O₈S₂
MDL Number: MFCD00007489
CAS Number: 3051-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR