2-(2-(Piperazin-1-yl)ethoxy)ethanol 95%

Supplier: Ambeed
Warning

Synonyms: 1-[2-(2-Hydroxyethoxy)ethyl]piperazine, 1-Hydroxyethylethoxypiperazine

A128169-100G A128169-10G A128169-25G A128169-1000G A128169-500G A128169-5G
76767-014EA 59 USD
76767-014 76767-016 76767-018 77624-080 77624-082 77624-084
2-(2-(Piperazin-1-yl)ethoxy)ethanol 95%
2-(2-(Piperazin-1-yl)ethoxy)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₂
MW: 174.2407 g/mol
MDL Number: MFCD00074931
CAS Number: 13349-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR