Ethyl-5-amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylic Acid Ethyl Ester, Ethyl 5-amino-4-cyano-3-(2-ethoxy-2-oxoethyl)thiophene-2-carboxylate

A128155-100G A128155-10G A128155-25G A128155-5G
76767-006EA 66 USD
76767-006 76767-008 76767-010 76767-012
Ethyl-5-amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate 97%
Ethyl-5-amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₄S
MDL Number: MFCD00832845
CAS Number: 58168-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR