3-Fluoro-5-(trifluoromethyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-(trifluoromethyl)benzeneboronic acid, (3-Fluoro-5-(trifluoromethyl)phenyl)boronic acid, 3-(Trifluoromethyl)-5-fluorophenylboronic acid

A128097-1G A128097-5G A128097-10G
76767-002EA 10 USD
76767-002 76767-004 77624-078
3-Fluoro-5-(trifluoromethyl)phenylboronic acid 97%
3-Fluoro-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₄O₂
MDL Number: MFCD06657735
CAS Number: 159020-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR