N-Carbobenzoxy-L-valine succinimidyl ester 95%

Supplier: Ambeed
Warning

Synonyms: N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, (S)-2,5-Dioxopyrrolidin-1-yl 2-(((benzyloxy)carbonyl)amino)-3-methylbutanoate, N-Cbz-L-valine Succinimidyl Ester, Z-Val-OSu

A127968-10G A127968-1G A127968-25G A127968-5G A127968-100G A127968-500G
76766-988EA 26.53 USD
76766-988 76766-990 76766-992 76766-994 77599-862 77624-076
N-Carbobenzoxy-L-valine succinimidyl ester 95%
N-Carbobenzoxy-L-valine succinimidyl ester

Precaución: for Research Use Only

Formula: C₁₇H₂₀N₂O₆
MW: 348.36 g/mol
MDL Number: MFCD00053547
CAS Number: 3496-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR