3-(Chloromethyl)-1H-1,2,4-triazol-5(4H)-one 97%
Supplier: Ambeed
|
Danger
|
A127880-1G
A127880-250MG
A127880-5G
76766-974EA
7.43
USD
76766-974
76766-976
76766-978
3-(Chloromethyl)-1H-1,2,4-triazol-5(4H)-one 97%
3-(Chloromethyl)-1H-1,2,4-triazol-5(4H)-one
Precaución: for Research Use Only
Formula:
C₃H₄ClN₃O MW: 133.54 g/mol |
MDL Number:
MFCD07776630 CAS Number: 252742-72-6 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...