4-Chloropyrimidin-5-amine 98%
Supplier: Ambeed
|
Warning
|
A127787-100MG
A127787-1G
A127787-250MG
A127787-5G
A127787-25G
76766-964EA
6
USD
76766-964
76766-966
76766-968
76766-970
77624-070
4-Chloropyrimidin-5-amine 98%
4-Chloropyrimidin-5-amine
Precaución: for Research Use Only
Formula:
C₄H₄ClN₃ |
MDL Number:
MFCD06412561 CAS Number: 54660-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...