5-Iodopyrimidin-2(1H)-one 95%

Supplier: Ambeed
Warning

A127274-1G A127274-250MG A127274-5G A127274-25G
76766-942EA 36.08 USD
76766-942 76766-944 77596-644 77624-062
5-Iodopyrimidin-2(1H)-one 95%
5-Iodopyrimidin-2(1H)-one

Precaución: for Research Use Only

Formula: C4H3IN2O
MW: 221.9800 g/mol
MDL Number: MFCD00130245
CAS Number: 79387-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR