5-Chloro-1,3-dihydro-2H-benzimidazol-2-one 98%

Supplier: Ambeed
Warning

A127264-1G A127264-250MG A127264-5G A127264-100MG
76766-936EA 14 USD
76766-936 76766-938 76766-940 77876-914
5-Chloro-1,3-dihydro-2H-benzimidazol-2-one 98%
5-Chloro-1,3-dihydro-2H-benzimidazol-2-one

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O
MDL Number: MFCD00493866
CAS Number: 2034-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR