5-Norbornene-2,3-dicarboxylic anhydride 95%

Supplier: Ambeed
Danger

A126691-100G A126691-10G A126691-25G A126691-500G
76766-922EA 10.61 USD
76766-922 76766-924 76766-926 76766-928
5-Norbornene-2,3-dicarboxylic anhydride 95%
5-Norbornene-2,3-dicarboxylic anhydride

Precaución: for Research Use Only

Formula: C₉H₈O₃
MDL Number: MFCD00069013
CAS Number: 826-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR