6-Methoxybenzo[d]thiazole 97%

Supplier: Ambeed
Warning

A126685-100MG A126685-10G A126685-1G A126685-250MG A126685-25G A126685-5G
76766-910EA 8.49 USD
76766-910 76766-912 76766-914 76766-916 76766-918 76766-920
6-Methoxybenzo[d]thiazole 97%
6-Methoxybenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₈H₇NOS
MW: 165.2100 g/mol
MDL Number: MFCD01961206
CAS Number: 2942-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR